(5E)-2-Mercapto-5-(2-nitrobenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039532-500MG
101847-674EA 144.9 USD
101847-674
(5E)-2-Mercapto-5-(2-nitrobenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-2-Mercapto-5-(2-nitrobenzylidene)-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₆H₁₁N₃O₃S
MDL Number: MFCD12027997
CAS Number: 53865-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR