(5E)-5-(4-Chlorobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039531-500MG
101847-672EA 193.85 USD
101847-672
(5E)-5-(4-Chlorobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-(4-Chlorobenzylidene)-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₆H₁₁ClN₂OS
MDL Number: MFCD12027996
CAS Number: 15985-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR