(5E)-5-Benzylidene-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Supplier: Matrix Scientific

039527-500MG
101847-664EA 144.9 USD
101847-664
(5E)-5-Benzylidene-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one
(5E)-5-Benzylidene-2-mercapto-3-phenyl-3,5-dihydro-4H-imidazol-4-one

Hazard: Irritant

Formula: C₁₆H₁₂N₂OS
MDL Number: MFCD06762654
CAS Number: 145693-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR