4-Methyl-1-(piperidin-4-ylcarbonyl)piperidine
Supplier: Matrix Scientific
039526-500MG
101847-662EA
387.69
USD
101847-662
4-Methyl-1-(piperidin-4-ylcarbonyl)piperidine
4-Methyl-1-(piperidin-4-ylcarbonyl)piperidine
Hazard: Irritant
Formula:
C₁₂H₂₂N₂O |
MDL Number:
MFCD03372493 CAS Number: 496057-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...