[(5-Methyl-2-phenyl-1,3-oxazol-4-yl)methyl]amine

Supplier: Matrix Scientific

039522-500MG
101847-654EA 289.8 USD
101847-654
[(5-Methyl-2-phenyl-1,3-oxazol-4-yl)methyl]amine
[(5-Methyl-2-phenyl-1,3-oxazol-4-yl)methyl]amine

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MDL Number: MFCD08271882
CAS Number: 132451-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR