3-(5-Methyl-1H-imidazol-1-yl)propanoic acid

Supplier: Matrix Scientific

039520-500MG
101847-650EA 287.5 USD
101847-650
3-(5-Methyl-1H-imidazol-1-yl)propanoic acid
3-(5-Methyl-1H-imidazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MDL Number: MFCD11053154
CAS Number: 1048920-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR