Ethyl-2-mercapto-4-phenyl-1,3-thiazole-5-carboxylate

Supplier: Matrix Scientific

039519-500MG
101847-648EA 290.77 USD
101847-648
Ethyl-2-mercapto-4-phenyl-1,3-thiazole-5-carboxylate
Ethyl-2-mercapto-4-phenyl-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₂S₂
MDL Number: MFCD12027992
CAS Number: 99822-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR