[2-(Isopropylthio)-4-oxo-4,5-dihydro-1,3-thiazol-5-yl]acetic acid

Supplier: Matrix Scientific

039514-500MG
101847-642EA 217.35 USD
101847-642
[2-(Isopropylthio)-4-oxo-4,5-dihydro-1,3-thiazol-5-yl]acetic acid
[2-(Isopropylthio)-4-oxo-4,5-dihydro-1,3-thiazol-5-yl]acetic acid

Hazard: Irritant

Formula: C₈H₁₁NO₃S₂
MDL Number: MFCD12027987
CAS Number: 1142201-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR