[1-(4-Methylphenyl)-1H-tetrazol-5-yl]methylimidothiocarbamate
Supplier: Matrix Scientific
039508-500MG
101847-630EA
290.77
USD
101847-630
[1-(4-Methylphenyl)-1H-tetrazol-5-yl]methylimidothiocarbamate
[1-(4-Methylphenyl)-1H-tetrazol-5-yl]methylimidothiocarbamate
Hazard: Irritant
Formula:
C₁₀H₁₂N₆S |
MDL Number:
MFCD12027983 CAS Number: 1142201-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...