5-(Chloromethyl)-1-phenyl-1H-tetrazole

Supplier: Matrix Scientific

039507-500MG
101847-628EA 287.5 USD
101847-628
5-(Chloromethyl)-1-phenyl-1H-tetrazole
5-(Chloromethyl)-1-phenyl-1H-tetrazole

Hazard: Irritant

Formula: C₈H₇ClN₄
MDL Number: MFCD03854522
CAS Number: 64473-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR