6-Fluoro-3-formylchromone

Supplier: Matrix Scientific

Synonyms: 6-Fluorochromone-3-carboxaldehyde6-Fluoro-4-oxo-4H-chromene-3-carbaldehyde

039506-500MG
101847-626EA 144.9 USD
101847-626
6-Fluoro-3-formylchromone
6-Fluoro-3-formylchromone

Hazard: Irritant

Formula: C₁₀H₅FO₃
MW: 192.15 g/mol
MDL Number: MFCD00139060
CAS Number: 69155-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR