8-(1,3-Benzothiazol-2-yl)-1-naphthoic acid

Supplier: Matrix Scientific

039505-500MG
101847-624EA 144.9 USD
101847-624
8-(1,3-Benzothiazol-2-yl)-1-naphthoic acid
8-(1,3-Benzothiazol-2-yl)-1-naphthoic acid

Hazard: Irritant

Formula: C₁₈H₁₁NO₂S
MDL Number: MFCD12027982
CAS Number: 1142201-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR