Methyl [(4-amino-1,3,5-triazin-2-yl)thio]acetate

Supplier: Matrix Scientific

039501-500MG
101847-616EA 290.77 USD
101847-616
Methyl [(4-amino-1,3,5-triazin-2-yl)thio]acetate
Methyl [(4-amino-1,3,5-triazin-2-yl)thio]acetate

Hazard: Irritant

Formula: C₆H₈N₄O₂S
MDL Number: MFCD12027979
CAS Number: 1142201-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR