4-(Methylthio)-1,3,5-triazin-2-amine
Supplier: Matrix Scientific
039492-500MG
101847-598EA
290.77
USD
101847-598
4-(Methylthio)-1,3,5-triazin-2-amine
4-(Methylthio)-1,3,5-triazin-2-amine
Hazard: Irritant
Formula:
C₄H₆N₄S |
MDL Number:
MFCD09971319 CAS Number: 27282-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...