4-(4-Amino-6-mercapto-1,2-dihydro-1,3,5-triazin-2-yl)-2-chlorophenol

Supplier: Matrix Scientific

039489-500MG
101847-592EA 144.9 USD
101847-592
4-(4-Amino-6-mercapto-1,2-dihydro-1,3,5-triazin-2-yl)-2-chlorophenol
4-(4-Amino-6-mercapto-1,2-dihydro-1,3,5-triazin-2-yl)-2-chlorophenol

Hazard: Irritant

Formula: C₉H₉ClN₄OS
MDL Number: MFCD12027973
CAS Number: 1142201-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR