4-Amino-6-(2,3,4-trimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039488-500MG
101847-590EA 144.9 USD
101847-590
4-Amino-6-(2,3,4-trimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2,3,4-trimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₃S
MDL Number: MFCD12027972
CAS Number: 1142208-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR