4-Amino-6-(5-methyl-2-furyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039475-500MG
101847-564EA 217.35 USD
101847-564
4-Amino-6-(5-methyl-2-furyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(5-methyl-2-furyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₈H₁₀N₄OS
MDL Number: MFCD12027959
CAS Number: 1142208-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR