4-Amino-6-(2,1,3-benzothiadiazol-4-yl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039474-500MG
101847-562EA 290.77 USD
101847-562
4-Amino-6-(2,1,3-benzothiadiazol-4-yl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2,1,3-benzothiadiazol-4-yl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₉H₈N₆S₂
MDL Number: MFCD12027958
CAS Number: 1142208-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR