4-Amino-6-(5-methoxy-1H-indol-3-yl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039471-500MG
101847-556EA 217.35 USD
101847-556
4-Amino-6-(5-methoxy-1H-indol-3-yl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(5-methoxy-1H-indol-3-yl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₂H₁₃N₅OS
MDL Number: MFCD12027955
CAS Number: 1142208-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR