4-Amino-6-(1H-indol-3-yl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039470-500MG
101847-554EA 217.35 USD
101847-554
4-Amino-6-(1H-indol-3-yl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(1H-indol-3-yl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₁H₁₁N₅S
MDL Number: MFCD12027954
CAS Number: 931302-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR