4-(4-Amino-6-mercapto-1,2-dihydro-1,3,5-triazin-2-yl)-3-methoxyphenol

Supplier: Matrix Scientific

039469-500MG
101847-552EA 217.35 USD
101847-552
4-(4-Amino-6-mercapto-1,2-dihydro-1,3,5-triazin-2-yl)-3-methoxyphenol
4-(4-Amino-6-mercapto-1,2-dihydro-1,3,5-triazin-2-yl)-3-methoxyphenol

Hazard: Irritant

Formula: C₁₀H₁₂N₄O₂S
MDL Number: MFCD12027953
CAS Number: 1142208-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR