4-Amino-6-(3-bromophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039467-500MG
101847-548EA 144.9 USD
101847-548
4-Amino-6-(3-bromophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(3-bromophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₉H₉BrN₄S
MDL Number: MFCD12027951
CAS Number: 1142208-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR