4-Amino-6-(3,4,5-trimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039465-500MG
101847-544EA 144.9 USD
101847-544
4-Amino-6-(3,4,5-trimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(3,4,5-trimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₃S
MDL Number: MFCD12027949
CAS Number: 1142208-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR