4-Amino-6-(4-methylphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
Supplier: Matrix Scientific
039463-500MG
101847-540EA
290.77
USD
101847-540
4-Amino-6-(4-methylphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(4-methylphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
Hazard: Irritant
Formula:
C₁₀H₁₂N₄S |
MDL Number:
MFCD12027947 CAS Number: 1142208-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...