4-Amino-6-(3-methylphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039462-500MG
101847-538EA 217.35 USD
101847-538
4-Amino-6-(3-methylphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(3-methylphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₀H₁₂N₄S
MDL Number: MFCD12027946
CAS Number: 1142208-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR