4-Amino-6-(2-thienyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039460-500MG
101847-534EA 217.35 USD
101847-534
4-Amino-6-(2-thienyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2-thienyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₇H₈N₄S₂
MDL Number: MFCD12027944
CAS Number: 1142208-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR