4-Amino-6-biphenyl-4-yl-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039459-500MG
101847-532EA 193.85 USD
101847-532
4-Amino-6-biphenyl-4-yl-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-biphenyl-4-yl-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₅H₁₄N₄S
MDL Number: MFCD12027943
CAS Number: 1142208-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR