4-Amino-6-(4-nitrophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039456-500MG
101847-526EA 217.35 USD
101847-526
4-Amino-6-(4-nitrophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(4-nitrophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₉H₉N₅O₂S
MDL Number: MFCD12027940
CAS Number: 1142208-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR