4-Amino-6-(4-fluorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
Supplier: Matrix Scientific
039452-500MG
101847-518EA
290.77
USD
101847-518
4-Amino-6-(4-fluorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(4-fluorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
Hazard: Irritant
Formula:
C₉H₉FN₄S |
MDL Number:
MFCD12027936 CAS Number: 1142207-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...