4-Amino-6-(4-ethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039450-500MG
101847-514EA 217.35 USD
101847-514
4-Amino-6-(4-ethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(4-ethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₁H₁₄N₄OS
MDL Number: MFCD12027934
CAS Number: 1142207-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR