4-Amino-6-(2-methoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039447-500MG
101847-508EA 217.35 USD
101847-508
4-Amino-6-(2-methoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2-methoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₀H₁₂N₄OS
MDL Number: MFCD12027931
CAS Number: 1142207-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR