4-Amino-6-[4-(dimethylamino)phenyl]-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039445-500MG
101847-504EA 290.77 USD
101847-504
4-Amino-6-[4-(dimethylamino)phenyl]-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-[4-(dimethylamino)phenyl]-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₁H₁₅N₅S
MDL Number: MFCD12027929
CAS Number: 1142207-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR