4-Amino-6-(2,5-dimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039443-500MG
101847-500EA 290.77 USD
101847-500
4-Amino-6-(2,5-dimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2,5-dimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₁H₁₄N₄O₂S
MDL Number: MFCD12027927
CAS Number: 1142207-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR