4-Amino-6-(2,4-dimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039442-500MG
101847-498EA 217.35 USD
101847-498
4-Amino-6-(2,4-dimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2,4-dimethoxyphenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₁₁H₁₄N₄O₂S
MDL Number: MFCD12027926
CAS Number: 1142207-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR