4-Amino-6-(2,6-dichlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
Supplier: Matrix Scientific
039440-500MG
101847-494EA
290.77
USD
101847-494
4-Amino-6-(2,6-dichlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2,6-dichlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
Hazard: Irritant
Formula:
C₉H₈Cl₂N₄S |
MDL Number:
MFCD12027924 CAS Number: 1142207-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...