4-Amino-6-(2,3-dichlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039438-500MG
101847-490EA 217.35 USD
101847-490
4-Amino-6-(2,3-dichlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2,3-dichlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₉H₈Cl₂N₄S
MDL Number: MFCD12027922
CAS Number: 1142207-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR