4-Amino-6-(4-chlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039436-500MG
101847-486EA 290.77 USD
101847-486
4-Amino-6-(4-chlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(4-chlorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₉H₉ClN₄S
MDL Number: MFCD12027920
CAS Number: 1142207-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR