4-Amino-6-(2-chloro-6-fluorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Supplier: Matrix Scientific

039433-500MG
101847-480EA 290.77 USD
101847-480
4-Amino-6-(2-chloro-6-fluorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol
4-Amino-6-(2-chloro-6-fluorophenyl)-1,6-dihydro-1,3,5-triazine-2-thiol

Hazard: Irritant

Formula: C₉H₈ClFN₄S
MDL Number: MFCD12027918
CAS Number: 1142207-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR