N-1,3-Benzodioxol-5-yl-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039425-500MG
101847-464EA 144.9 USD
101847-464
N-1,3-Benzodioxol-5-yl-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-1,3-Benzodioxol-5-yl-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₄S₂
MDL Number: MFCD12027910
CAS Number: 1142207-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR