N-[4-(1,3-Benzoxazol-2-yl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039422-500MG
101847-458EA 144.9 USD
101847-458
N-[4-(1,3-Benzoxazol-2-yl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-[4-(1,3-Benzoxazol-2-yl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₈H₁₃N₃O₃S₂
MDL Number: MFCD12027907
CAS Number: 1142207-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR