N-[4-(Aminosulfonyl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039415-500MG
101847-446EA 193.85 USD
101847-446
N-[4-(Aminosulfonyl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-[4-(Aminosulfonyl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₁N₃O₄S₃
MDL Number: MFCD12027900
CAS Number: 1142207-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR