2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(2-methyl-5-nitrophenyl)acetamide

Supplier: Matrix Scientific

039403-500MG
101847-422EA 144.9 USD
101847-422
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(2-methyl-5-nitrophenyl)acetamide
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(2-methyl-5-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₄S₂
MDL Number: MFCD12027888
CAS Number: 1142206-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR