N-(4-Fluorophenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039399-500MG
101847-414EA 193.85 USD
101847-414
N-(4-Fluorophenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-(4-Fluorophenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₁H₉FN₂O₂S₂
MDL Number: MFCD12027884
CAS Number: 1142206-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR