2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-methoxyphenyl)acetamide

Supplier: Matrix Scientific

039396-500MG
101847-408EA 193.85 USD
101847-408
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-methoxyphenyl)acetamide
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-methoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₃S₂
MDL Number: MFCD12027881
CAS Number: 1142206-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR