N-(4-Acetylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039386-500MG
101847-388EA 193.85 USD
101847-388
N-(4-Acetylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-(4-Acetylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₃S₂
MDL Number: MFCD12027871
CAS Number: 1142206-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR