N-[2-Chloro-5-(trifluoromethyl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039382-500MG
101847-380EA 193.85 USD
101847-380
N-[2-Chloro-5-(trifluoromethyl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-[2-Chloro-5-(trifluoromethyl)phenyl]-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₂H₈ClF₃N₂O₂S₂
MDL Number: MFCD12027867
CAS Number: 1142200-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR