2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-methylphenyl)acetamide

Supplier: Matrix Scientific

039380-500MG
101847-376EA 193.85 USD
101847-376
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-methylphenyl)acetamide
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-methylphenyl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S₂
MDL Number: MFCD12027865
CAS Number: 1142200-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR