2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-nitrophenyl)acetamide

Supplier: Matrix Scientific

039377-500MG
101847-370EA 193.85 USD
101847-370
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-nitrophenyl)acetamide
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-(4-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₁₁H₉N₃O₄S₂
MDL Number: MFCD12027862
CAS Number: 1142200-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR