N-(3,5-Dimethylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
Supplier: Matrix Scientific
039376-500MG
101847-368EA
193.85
USD
101847-368
N-(3,5-Dimethylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-(3,5-Dimethylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₂S₂ |
MDL Number:
MFCD12027861 CAS Number: 1142200-40-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...