N-(3,4-Dimethylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039375-500MG
101847-366EA 144.9 USD
101847-366
N-(3,4-Dimethylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-(3,4-Dimethylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂S₂
MDL Number: MFCD12027860
CAS Number: 1142200-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR